Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGACCCCAAGGGCTGCAAAGCAT[A/T]GCTCACCTTCAGCTTCCGCTTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611508 MIM: 131370 MIM: 610056 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMTA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMTA2 - calmodulin binding transcription activator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ENO3 - enolase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6864 - microRNA 6864 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPAG7 - sperm associated antigen 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004890.2 | Intron | NP_004881.2 | ||||
XM_006721600.2 | Intron | XP_006721663.1 |