Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACGACAGGCAATCAGACAAGGAT[A/G]GACCTCCAGCTACAAGCAGCACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AURKB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AURKB - aurora kinase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256834.2 | Intron | NP_001243763.1 | ||||
NM_001284526.1 | Intron | NP_001271455.1 | ||||
NM_001313950.1 | Intron | NP_001300879.1 | ||||
NM_001313951.1 | Intron | NP_001300880.1 | ||||
NM_001313952.1 | Intron | NP_001300881.1 | ||||
NM_001313953.1 | Intron | NP_001300882.1 | ||||
NM_001313954.1 | Intron | NP_001300883.1 | ||||
NM_001313955.1 | Intron | NP_001300884.1 | ||||
NM_004217.3 | Intron | NP_004208.2 | ||||
XM_011524070.2 | Intron | XP_011522372.1 | ||||
XM_011524072.2 | Intron | XP_011522374.1 | ||||
XM_017025307.1 | Intron | XP_016880796.1 | ||||
XM_017025308.1 | Intron | XP_016880797.1 | ||||
XM_017025309.1 | Intron | XP_016880798.1 | ||||
XM_017025310.1 | Intron | XP_016880799.1 | ||||
XM_017025311.1 | Intron | XP_016880800.1 |