Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTCAGATGACGAGGACGATGA[C/T]GAGGAGGAATATGGCAAGGAGGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601333 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUPT6H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUPT6H - SPT6 homolog, histone chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320755.1 | 671 | Silent Mutation | GAC,GAT | D,D 131 | NP_001307684.1 | |
NM_003170.4 | 671 | Silent Mutation | GAC,GAT | D,D 131 | NP_003161.2 | |
XM_017024978.1 | 671 | Silent Mutation | GAC,GAT | D,D 131 | XP_016880467.1 | |
XM_017024979.1 | 671 | Silent Mutation | GAC,GAT | D,D 131 | XP_016880468.1 | |
XM_017024980.1 | 671 | Silent Mutation | GAC,GAT | D,D 8 | XP_016880469.1 |