Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTCCTACTCCATCTCCAAGCCT[A/C]ACCCTAGCATCTTCTCTGGCCCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147910 MIM: 610601 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK1 - kallikrein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLK15 - kallikrein related peptidase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277081.1 | Intron | NP_001264010.1 | ||||
NM_001277082.1 | Intron | NP_001264011.1 | ||||
NM_017509.3 | Intron | NP_059979.2 | ||||
XM_006723265.3 | Intron | XP_006723328.1 | ||||
XM_011527085.2 | Intron | XP_011525387.1 | ||||
XM_011527087.2 | Intron | XP_011525389.1 | ||||
XM_011527088.2 | Intron | XP_011525390.1 | ||||
XM_011527089.2 | Intron | XP_011525391.1 | ||||
XM_017026943.1 | Intron | XP_016882432.1 |
LOC105372441 - uncharacterized LOC105372441 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MGC45922 - uncharacterized LOC284365 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |