Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCATCCAGTGCCGGGTGCTGCC[C/G]CCCAGCCACCGCGTGACCTGGGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604721 MIM: 164875 | ||||||||||||||||||||
Literature Links: |
SH2D3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SH2D3A - SH2 domain containing 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAV1 - vav guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258206.1 | 194 | Silent Mutation | CCC,CCG | P,P 18 | NP_001245135.1 | |
NM_001258207.1 | 194 | Silent Mutation | CCC,CCG | P,P 18 | NP_001245136.1 | |
NM_005428.3 | 194 | Silent Mutation | CCC,CCG | P,P 18 | NP_005419.2 | |
XM_005259642.1 | 194 | Silent Mutation | CCC,CCG | P,P 18 | XP_005259699.1 |