Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCTGGGCCTGGCCAAGGCGGGC[A/T]TGGAGGCCGCGCTGCTCAACGTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600691 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC27A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC27A1 - solute carrier family 27 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198580.2 | 494 | Missense Mutation | ATG,TTG | M,L 153 | NP_940982.1 | |
XM_011528000.1 | 494 | Missense Mutation | ATG,TTG | M,L 153 | XP_011526302.1 | |
XM_011528001.2 | 494 | Missense Mutation | ATG,TTG | M,L 153 | XP_011526303.1 | |
XM_011528002.2 | 494 | Missense Mutation | ATG,TTG | M,L 153 | XP_011526304.1 | |
XM_011528003.2 | 494 | Missense Mutation | ATG,TTG | M,L 153 | XP_011526305.1 | |
XM_017026781.1 | 494 | Missense Mutation | ATG,TTG | M,L 153 | XP_016882270.1 |