Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCGGGGGAGGACGAAGAGAGAC[G/T]AGAGCGGAGCCCGGAGGGGCCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615321 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLIC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLIC6 - chloride intracellular channel 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317009.1 | 1557 | Missense Mutation | CGA,CTA | R,L 374 | NP_001303938.1 | |
NM_053277.2 | 1557 | Missense Mutation | CGA,CTA | R,L 374 | NP_444507.1 | |
XM_017028406.1 | 1557 | Missense Mutation | CGA,CTA | R,L 374 | XP_016883895.1 |