Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAAAGGTGGAAGAGGAAGATGA[A/G]GAAGAGGAGGAGGAAGAAGAGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300193 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGB3 - high mobility group box 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301228.1 | 662 | Silent Mutation | GAA,GAG | E,E 185 | NP_001288157.1 | |
NM_001301229.1 | 662 | Silent Mutation | GAA,GAG | E,E 185 | NP_001288158.1 | |
NM_001301231.1 | 662 | Silent Mutation | GAA,GAG | E,E 205 | NP_001288160.1 | |
NM_005342.3 | 662 | Silent Mutation | GAA,GAG | E,E 185 | NP_005333.2 |