Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAGCGACCGCTTCCGGTCCTT[A/C]GATGCGCTCCTCATAGAGCTCACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613166 | ||||||||||||||||||||
Literature Links: |
DCLK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCLK2 - doublecortin like kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040260.3 | 1051 | Missense Mutation | TTA,TTC | L,F 99 | NP_001035350.2 | |
NM_001040261.4 | 1051 | Missense Mutation | TTA,TTC | L,F 99 | NP_001035351.4 | |
XM_005262785.2 | 1051 | Missense Mutation | TTA,TTC | L,F 99 | XP_005262842.1 | |
XM_005262786.2 | 1051 | Missense Mutation | TTA,TTC | L,F 99 | XP_005262843.1 | |
XM_017007832.1 | 1051 | Missense Mutation | TTA,TTC | L,F 99 | XP_016863321.1 | |
XM_017007833.1 | 1051 | Missense Mutation | TTA,TTC | L,F 99 | XP_016863322.1 |