Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTTTTTTTAATACCATTCT[A/G]TGATTTAAACTTTTCTATAATGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPRKB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TPRKB - TP53RK binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005264343.3 | Intron | XP_005264400.1 | ||||
XM_006712026.3 | Intron | XP_006712089.1 | ||||
XM_006712027.3 | Intron | XP_006712090.1 | ||||
XM_011532877.2 | Intron | XP_011531179.1 | ||||
XM_011532878.2 | Intron | XP_011531180.1 | ||||
XM_011532879.2 | Intron | XP_011531181.1 | ||||
XM_011532880.2 | Intron | XP_011531182.1 | ||||
XM_017004232.1 | Intron | XP_016859721.1 | ||||
XM_017004233.1 | Intron | XP_016859722.1 | ||||
XM_017004234.1 | Intron | XP_016859723.1 | ||||
XM_017004235.1 | Intron | XP_016859724.1 |