Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGCCCCAGCCTCAGCCCCTCC[A/G]GGCCCCAGTGAGGAGCTGCTGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130660 MIM: 614058 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMILIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMILIN1 - elastin microfibril interfacer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007046.3 | 1351 | Silent Mutation | CCA,CCG | P,P 284 | NP_008977.1 |
KHK - ketohexokinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |