Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTTTGTGGGCAACATGCTGGTC[A/G]TCCTCATCTTAATAAACTGCAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601267 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCR2 - C-C motif chemokine receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123041.2 | 388 | Missense Mutation | ATC,GTC | I,V 64 | NP_001116513.2 | |
NM_001123396.1 | 388 | Missense Mutation | ATC,GTC | I,V 64 | NP_001116868.1 | |
XM_011534069.1 | 388 | Missense Mutation | ATC,GTC | I,V 64 | XP_011532371.1 |
LOC102724297 - uncharacterized LOC102724297 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |