Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCCGGAGCAGGGGAGCAGAGGC[C/G]GAGAGCAGCCAAAGCGCCCAATGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610011 MIM: 614589 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARSK - arylsulfatase family member K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198150.2 | 235 | Missense Mutation | CCG,CGG | P,R 26 | NP_937793.1 | |
XM_005271904.3 | 235 | UTR 5 | XP_005271961.1 |
TTC37 - tetratricopeptide repeat domain 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014639.3 | 235 | Intron | NP_055454.1 |