Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGGTGGTGGTGGTGGTGGTGGT[A/G]GTTCTTTGAGAGAAGCTTTGCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602866 MIM: 601735 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASIC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASIC1 - acid sensing ion channel subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMARCD1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003076.4 | Intron | NP_003067.3 | ||||
NM_139071.2 | Intron | NP_620710.2 | ||||
XM_005269107.3 | Intron | XP_005269164.2 | ||||
XM_011538695.1 | Intron | XP_011536997.1 |