Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAGAAGGAGAAGGTCTGAGCCAG[A/G]ATGAGACGGTCCTCCTGGGCGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615126 MIM: 606780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD13D - ankyrin repeat domain 13D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSH3 - slingshot protein phosphatase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017857.3 | 602 | Missense Mutation | AAT,GAT | N,D 142 | NP_060327.3 | |
XM_017017942.1 | 602 | Missense Mutation | AAT,GAT | N,D 42 | XP_016873431.1 |