Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603843 MIM: 603624 MIM: 605915 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFB10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFB10 - NADH:ubiquinone oxidoreductase subunit B10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF151 - ring finger protein 151 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174903.4 | 636 | Silent Mutation | GTG,GTT | V,V 122 | NP_777563.2 | |
XM_005255129.4 | 636 | Silent Mutation | GTG,GTT | V,V 131 | XP_005255186.1 |
RPS2 - ribosomal protein S2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNHG9 - small nucleolar RNA host gene 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA10 - small nucleolar RNA, H/ACA box 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA64 - small nucleolar RNA, H/ACA box 64 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA78 - small nucleolar RNA, H/ACA box 78 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBL3 - transducin beta like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |