Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCCCAACAGCACGGCGTGGCCT[C/T]TCAGCCTCGGTGAGTACCCGCCGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 110750 | ||||||||||||||||||||
Literature Links: |
GYPC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GYPC - glycophorin C (Gerbich blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256584.1 | 369 | UTR 5 | NP_001243513.1 | |||
NM_002101.4 | 369 | Missense Mutation | CTC,TTC | L,F 14 | NP_002092.1 | |
NM_016815.3 | 369 | Missense Mutation | CTC,TTC | L,F 14 | NP_058131.1 | |
XM_006712460.3 | 369 | Intron | XP_006712523.1 |