Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGTCCCCAAAGGCCTGAAAAGTC[C/G]ACCAGAGCCATGGGGCTGGCCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124060 | ||||||||||||||||||||||||||||||||
Literature Links: |
CYP1A2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
CYP1A2*15,g.125C>G | ||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CYP1A2 - cytochrome P450 family 1 subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000761.4 | 189 | Missense Mutation | CCA,CGA | P,R 42 | NP_000752.2 |