Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCATCTCTAGTCTTTGCTAAG[C/T]GAGAGTCACTCACAACTCTGCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601883 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf174 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf174 - chromosome 1 open reading frame 174 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207356.2 | 2987 | Missense Mutation | CAC,CGC | H,R 131 | NP_997239.2 | |
XM_005244743.3 | 2987 | Missense Mutation | CAC,CGC | H,R 142 | XP_005244800.1 | |
XM_011541323.2 | 2987 | Missense Mutation | CAC,CGC | H,R 142 | XP_011539625.1 |
DFFB - DNA fragmentation factor subunit beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC01134 - long intergenic non-protein coding RNA 1134 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |