Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTCACTTGGTCTTTGCATCCCA[C/T]GCCTCATGTTACCTAAGGAGGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC27 - coiled-coil domain containing 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152492.2 | 211 | Missense Mutation | CGC,TGC | R,C 43 | NP_689705.2 | |
XM_011540780.1 | 211 | Missense Mutation | CGC,TGC | R,C 43 | XP_011539082.1 | |
XM_017000402.1 | 211 | Missense Mutation | CGC,TGC | R,C 43 | XP_016855891.1 | |
XM_017000403.1 | 211 | Missense Mutation | CGC,TGC | R,C 43 | XP_016855892.1 | |
XM_017000404.1 | 211 | Missense Mutation | CGC,TGC | R,C 43 | XP_016855893.1 | |
XM_017000405.1 | 211 | Missense Mutation | CGC,TGC | R,C 43 | XP_016855894.1 | |
XM_017000406.1 | 211 | Intron | XP_016855895.1 |
TP73-AS1 - TP73 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |