Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACATTTTTAAAAACTGGCACAC[C/T]TTGGTCTGATACAAGATCCTGTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604513 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD84 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD84 - CD84 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184879.1 | 4087 | UTR 3 | NP_001171808.1 | |||
NM_001184881.1 | 4087 | UTR 3 | NP_001171810.1 | |||
NM_001184882.1 | 4087 | UTR 3 | NP_001171811.1 | |||
NM_003874.3 | 4087 | UTR 3 | NP_003865.1 | |||
XM_011510094.2 | 4087 | UTR 3 | XP_011508396.1 | |||
XM_011510095.2 | 4087 | UTR 3 | XP_011508397.1 |