Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTTGGCCCTGGTCTCCTGCCT[C/T]TTTCTGTGCTCTCTGGTCTGGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUCO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUCO - SUN domain containing ossification factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282750.1 | 441 | Silent Mutation | CTC,CTT | L,L 14 | NP_001269679.1 | |
NM_001282751.1 | 441 | UTR 5 | NP_001269680.1 | |||
NM_014283.4 | 441 | Silent Mutation | CTC,CTT | L,L 14 | NP_055098.1 | |
NM_016227.3 | 441 | Silent Mutation | CTC,CTT | L,L 209 | NP_057311.3 | |
XM_006711374.2 | 441 | Silent Mutation | CTC,CTT | L,L 14 | XP_006711437.1 | |
XM_006711375.2 | 441 | Silent Mutation | CTC,CTT | L,L 14 | XP_006711438.1 | |
XM_006711376.1 | 441 | Silent Mutation | CTC,CTT | L,L 14 | XP_006711439.1 |