Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGCGACACCGACAGTGGCATCA[A/T]CCTGCAGTCTGGTGAGTGTTCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf106 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf106 - chromosome 1 open reading frame 106 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142569.2 | 405 | Missense Mutation | AAC,ATC | N,I 15 | NP_001136041.1 | |
NM_018265.3 | 405 | Missense Mutation | AAC,ATC | N,I 114 | NP_060735.3 | |
XM_011509754.2 | 405 | Missense Mutation | AAC,ATC | N,I 15 | XP_011508056.1 | |
XM_011509755.1 | 405 | Missense Mutation | AAC,ATC | N,I 15 | XP_011508057.1 | |
XM_011509756.2 | 405 | Missense Mutation | AAC,ATC | N,I 114 | XP_011508058.1 |