Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAGAAAGCAGGTGAGAGGGGTA[A/G]CTCCTTGGTCAGCTGCCCAGCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607622 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PBXIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PBXIP1 - PBX homeobox interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317734.1 | 1617 | Silent Mutation | CTA,TTA | L,L 607 | NP_001304663.1 | |
NM_001317735.1 | 1617 | Silent Mutation | CTA,TTA | L,L 481 | NP_001304664.1 | |
NM_020524.3 | 1617 | Silent Mutation | CTA,TTA | L,L 636 | NP_065385.2 |
PMVK - phosphomevalonate kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |