Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTATTCCTGAGTTCATACCAACA[A/C]CTGCTAGCTCTCCCCTCTAGCGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164873 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYCSP52 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYCSP52 - cytochrome c, somatic pseudogene 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ETV3 - ETS variant 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145312.1 | 4005 | Intron | NP_001138784.1 | |||
NM_005240.1 | 4005 | Intron | NP_005231.1 | |||
XM_006711210.2 | 4005 | UTR 3 | XP_006711273.1 |