Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGCACACGCCTGAAAAGTGGGT[A/G]AGGTTCAAGTACCCAAAGCTCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 132810 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPHX1 - epoxide hydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000120.3 | 1488 | Silent Mutation | GTA,GTG | V,V 413 | NP_000111.1 | |
NM_001136018.3 | 1488 | Silent Mutation | GTA,GTG | V,V 413 | NP_001129490.1 | |
NM_001291163.1 | 1488 | Silent Mutation | GTA,GTG | V,V 413 | NP_001278092.1 |
TMEM63A - transmembrane protein 63A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014698.2 | 1488 | Intron | NP_055513.2 | |||
XM_006711841.3 | 1488 | Intron | XP_006711904.1 | |||
XM_011544328.2 | 1488 | Intron | XP_011542630.1 | |||
XM_011544329.2 | 1488 | Intron | XP_011542631.1 | |||
XM_011544330.2 | 1488 | Intron | XP_011542632.1 | |||
XM_011544331.2 | 1488 | Intron | XP_011542633.1 | |||
XM_011544332.2 | 1488 | Intron | XP_011542634.1 | |||
XM_017002916.1 | 1488 | Intron | XP_016858405.1 |