Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGGATCTCCTAAGAGCAGGAGT[A/G]AGCTGCCTTCTTTGTACCTGCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601109 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HTR6 - 5-hydroxytryptamine receptor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMCO4 - transmembrane and coiled-coil domains 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181719.4 | 3142 | UTR 3 | NP_859070.3 | |||
XM_005245822.4 | 3142 | UTR 3 | XP_005245879.1 | |||
XM_005245824.4 | 3142 | UTR 3 | XP_005245881.1 | |||
XM_005245825.4 | 3142 | UTR 3 | XP_005245882.1 | |||
XM_005245827.3 | 3142 | UTR 3 | XP_005245884.1 | |||
XM_006710551.3 | 3142 | UTR 3 | XP_006710614.1 | |||
XM_011541179.2 | 3142 | UTR 3 | XP_011539481.1 | |||
XM_011541180.2 | 3142 | UTR 3 | XP_011539482.1 | |||
XM_011541181.1 | 3142 | UTR 3 | XP_011539483.1 | |||
XM_011541185.2 | 3142 | UTR 3 | XP_011539487.1 | |||
XM_011541186.2 | 3142 | UTR 3 | XP_011539488.1 | |||
XM_017000913.1 | 3142 | UTR 3 | XP_016856402.1 | |||
XM_017000914.1 | 3142 | UTR 3 | XP_016856403.1 | |||
XM_017000915.1 | 3142 | UTR 3 | XP_016856404.1 | |||
XM_017000916.1 | 3142 | UTR 3 | XP_016856405.1 | |||
XM_017000917.1 | 3142 | UTR 3 | XP_016856406.1 | |||
XM_017000918.1 | 3142 | UTR 3 | XP_016856407.1 | |||
XM_017000919.1 | 3142 | UTR 3 | XP_016856408.1 | |||
XM_017000920.1 | 3142 | UTR 3 | XP_016856409.1 | |||
XM_017000921.1 | 3142 | UTR 3 | XP_016856410.1 | |||
XM_017000922.1 | 3142 | UTR 3 | XP_016856411.1 |