Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGGTTCTGGTGTTGCTTTGGGC[A/G]CCAGGGATGCAACTGAGTTTAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605630 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G2D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G2D - phospholipase A2 group IID | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271814.1 | 1072 | UTR 3 | NP_001258743.1 | |||
NM_012400.3 | 1072 | UTR 3 | NP_036532.1 |