Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCTTTTATATTGGCCTGAGAGG[A/C]GAGTGGACTGAGGTAAGATGGGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606953 MIM: 616143 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALE - UDP-galactose-4-epimerase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYPLA2 - lysophospholipase II | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITHD1 - PITH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020362.4 | 577 | Silent Mutation | GGA,GGC | G,G 174 | NP_065095.2 | |
XM_011541804.1 | 577 | Silent Mutation | GGA,GGC | G,G 81 | XP_011540106.1 |
TCEB3-AS1 - TCEB3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |