Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCTCCCCTGCAGAGTACCATTC[A/G]CGAGTCCTACGGTCGGCCAGAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609121 MIM: 603272 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPER4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CATSPER4 - cation channel sperm associated 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198137.1 | 149 | Missense Mutation | CAC,CGC | H,R 50 | NP_937770.1 | |
XM_011541432.2 | 149 | Missense Mutation | CAC,CGC | H,R 50 | XP_011539734.1 | |
XM_011541433.2 | 149 | Missense Mutation | CAC,CGC | H,R 50 | XP_011539735.1 |
CNKSR1 - connector enhancer of kinase suppressor of Ras 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297647.1 | 149 | Intron | NP_001284576.1 | |||
NM_001297648.1 | 149 | Intron | NP_001284577.1 | |||
NM_006314.2 | 149 | Intron | NP_006305.2 | |||
XM_011540484.2 | 149 | Intron | XP_011538786.1 | |||
XM_011540485.2 | 149 | Intron | XP_011538787.1 |