Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCACAAAGTATATTTTCTCTATT[A/T]ATGAGTTCATCCAGTGGTTTTCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606435 MIM: 606433 | ||||||||||||||||||||
Literature Links: |
UGT1A10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UGT1A10 - UDP glucuronosyltransferase family 1 member A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019075.2 | 373 | Missense Mutation | TTA,TTT | L,F 109 | NP_061948.1 |
UGT1A8 - UDP glucuronosyltransferase family 1 member A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019076.4 | 373 | Intron | NP_061949.3 |