Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTAACTGATGTTTCTGAGTTAG[A/G]GAAAAATTCACTTGCTATCAACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138280 MIM: 600555 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLS - glutaminase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT1 - signal transducer and activator of transcription 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007315.3 | 2795 | UTR 3 | NP_009330.1 | |||
NM_139266.2 | 2795 | Intron | NP_644671.1 | |||
XM_006712718.1 | 2795 | UTR 3 | XP_006712781.1 | |||
XM_017004783.1 | 2795 | UTR 3 | XP_016860272.1 |