Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAGTCAAGGATTCCTCCATTCT[C/T]GCTTCATGAAATGAGCAGTTTTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606597 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAX3 - paired box 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000438.5 | 2339 | Intron | NP_000429.2 | |||
NM_001127366.2 | 2339 | UTR 3 | NP_001120838.1 | |||
NM_013942.4 | 2339 | Intron | NP_039230.1 | |||
NM_181457.3 | 2339 | Intron | NP_852122.1 | |||
NM_181458.3 | 2339 | UTR 3 | NP_852123.1 | |||
NM_181459.3 | 2339 | UTR 3 | NP_852124.1 | |||
NM_181460.3 | 2339 | UTR 3 | NP_852125.1 | |||
NM_181461.3 | 2339 | UTR 3 | NP_852126.1 |