Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAATGGCGGAAGGAGAAGATGTG[C/G]CGCCGCTGCCAACGTCGAGCGGCGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
TBC1D23 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TBC1D23 - TBC1 domain family member 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199198.2 | 56 | Missense Mutation | CCG,GCG | P,A 8 | NP_001186127.1 | |
NM_018309.4 | 56 | Missense Mutation | CCG,GCG | P,A 8 | NP_060779.2 | |
XM_011512974.2 | 56 | Intron | XP_011511276.1 | |||
XM_017006840.1 | 56 | Missense Mutation | CCG,GCG | P,A 8 | XP_016862329.1 | |
XM_017006841.1 | 56 | UTR 5 | XP_016862330.1 |