Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAATGTCTTCCGAACTCAACATA[A/G]TATTCTAACAGAATTGTAGGTAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611408 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCA5 - LCA5, lebercilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122769.2 | 3205 | UTR 3 | NP_001116241.1 | |||
NM_181714.3 | 3205 | UTR 3 | NP_859065.2 | |||
XM_005248665.4 | 3205 | UTR 3 | XP_005248722.1 | |||
XM_011535504.1 | 3205 | UTR 3 | XP_011533806.1 |