Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGGCCTGGTTAAACTGGAAAGAC[A/C]AGGAAAGCCCGCCCACGGGCGTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604092 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTK - TTK protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166691.1 | 277 | Intron | NP_001160163.1 | |||
NM_003318.4 | 277 | Intron | NP_003309.2 | |||
XM_011536099.2 | 277 | Intron | XP_011534401.1 | |||
XM_011536100.2 | 277 | Intron | XP_011534402.1 | |||
XM_017011242.1 | 277 | UTR 5 | XP_016866731.1 |