Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTCCGCAGGCTCACTCGGTCAG[T/C]CTGTGCCTGGCGCTTGTACTTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HLA-C - major histocompatibility complex, class I, C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243042.1 | 354 | Missense Mutation | ACT,GCT | T,A 97 | NP_001229971.1 | |
NM_002117.5 | 354 | Missense Mutation | ACT,GCT | T,A 97 | NP_002108.4 |