Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGTGCCAGCCCCCATCTGTCCAC[C/T]GCCATCTAGCCCCCAGCTCTGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613976 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FANCE - Fanconi anemia complementation group E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021922.2 | Intron | NP_068741.1 | ||||
XM_005248885.2 | Intron | XP_005248942.1 | ||||
XM_005248886.2 | Intron | XP_005248943.1 | ||||
XM_005248887.2 | Intron | XP_005248944.1 | ||||
XM_005248888.3 | Intron | XP_005248945.1 | ||||
XM_011514343.2 | Intron | XP_011512645.1 |