Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTTCTGCTCTTGTACATTTTAC[A/G]CATGCAGTGACTCTTGCCTGTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 112265 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMP5 - bone morphogenetic protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005249304.3 | 2825 | UTR 3 | XP_005249361.1 | |||
XM_011514816.2 | 2825 | Intron | XP_011513118.1 | |||
XM_011514817.2 | 2825 | Intron | XP_011513119.1 | |||
XM_017011198.1 | 2825 | UTR 3 | XP_016866687.1 |