Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGTGCAGCTTTTTTCTTTTGAA[C/T]GCTTTACTTTATAAAATATATAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171060 MIM: 606090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCB4 - ATP binding cassette subfamily B member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000443.3 | Intron | NP_000434.1 | ||||
NM_018849.2 | Intron | NP_061337.1 | ||||
NM_018850.2 | Intron | NP_061338.1 | ||||
XM_011516308.2 | Intron | XP_011514610.2 | ||||
XM_011516309.2 | Intron | XP_011514611.2 | ||||
XM_011516310.2 | Intron | XP_011514612.2 | ||||
XM_011516311.2 | Intron | XP_011514613.2 | ||||
XM_011516312.2 | Intron | XP_011514614.2 | ||||
XM_011516313.2 | Intron | XP_011514615.1 | ||||
XM_011516315.2 | Intron | XP_011514617.2 | ||||
XM_017012323.1 | Intron | XP_016867812.1 |
CROT - carnitine O-octanoyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |