Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATTTTTGAACTTTTAAAGTTCT[A/G]GTTTGGTCTTTGAATCAAAACAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606650 MIM: 613821 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIN3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIN3A - glutamate ionotropic receptor NMDA type subunit 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133445.2 | 884 | Intron | NP_597702.2 | |||
XM_011518211.2 | 884 | Intron | XP_011516513.1 |
PPP3R2 - protein phosphatase 3 regulatory subunit B, beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147180.3 | 884 | UTR 3 | NP_671709.1 |