Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTGCTGCCGCCGCCCTTGCCAA[C/T]GTTGTTAGCCCGGTGACAGCGAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612265 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM120A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM120A - family with sequence similarity 120A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286722.1 | 695 | Intron | NP_001273651.1 | |||
NM_001286723.1 | 695 | Intron | NP_001273652.1 | |||
NM_001286724.1 | 695 | Intron | NP_001273653.1 | |||
NM_014612.4 | 695 | Intron | NP_055427.2 | |||
XM_005251842.4 | 695 | Intron | XP_005251899.1 | |||
XM_011518412.2 | 695 | Intron | XP_011516714.1 | |||
XM_011518413.2 | 695 | Intron | XP_011516715.1 | |||
XM_011518414.2 | 695 | Intron | XP_011516716.1 | |||
XM_011518416.2 | 695 | Intron | XP_011516718.1 |
FAM120AOS - family with sequence similarity 120A opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322224.1 | 695 | Intron | NP_001309153.1 | |||
NM_198841.2 | 695 | UTR 5 | NP_942138.2 |