Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAACTACAATGATGCATTGTTGC[A/G]GTACTTTGATATAAAATCAGAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605491 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEBL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEBL - nebulette | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173484.1 | 8810 | UTR 3 | NP_001166955.1 | |||
NM_006393.2 | 8810 | UTR 3 | NP_006384.1 | |||
NM_213569.2 | 8810 | UTR 3 | NP_998734.1 | |||
XM_005252342.4 | 8810 | UTR 3 | XP_005252399.1 | |||
XM_005252343.4 | 8810 | UTR 3 | XP_005252400.1 | |||
XM_005252344.4 | 8810 | UTR 3 | XP_005252401.1 | |||
XM_011519291.2 | 8810 | UTR 3 | XP_011517593.1 | |||
XM_017015467.1 | 8810 | UTR 3 | XP_016870956.1 | |||
XM_017015468.1 | 8810 | UTR 3 | XP_016870957.1 | |||
XM_017015469.1 | 8810 | Intron | XP_016870958.1 |