Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCAGCTTGTAGCCAGCCGGGGC[A/G]AGTGGCACGTTTATTTAACTTTTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf43 - chromosome 12 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286191.1 | 2414 | UTR 3 | NP_001273120.1 | |||
NM_001286192.1 | 2414 | UTR 3 | NP_001273121.1 | |||
NM_001286195.1 | 2414 | UTR 3 | NP_001273124.1 | |||
NM_001286196.1 | 2414 | UTR 3 | NP_001273125.1 | |||
NM_001286197.1 | 2414 | UTR 3 | NP_001273126.1 | |||
NM_001286198.1 | 2414 | UTR 3 | NP_001273127.1 | |||
NM_022895.2 | 2414 | UTR 3 | NP_075046.1 | |||
XM_017019829.1 | 2414 | Intron | XP_016875318.1 |
HNF1A - HNF1 homeobox A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000545.6 | 2414 | UTR 3 | NP_000536.5 | |||
NM_001306179.1 | 2414 | UTR 3 | NP_001293108.1 | |||
XM_005253931.3 | 2414 | UTR 3 | XP_005253988.1 |