Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCCAAGGGCTGCCTCTGATGGC[A/G]GACGGGGGTGTGGTCCTGGGACTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176872 MIM: 605979 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP2K1 - mitogen-activated protein kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002755.3 | Intron | NP_002746.1 | ||||
XM_011521783.2 | Intron | XP_011520085.1 | ||||
XM_017022411.1 | Intron | XP_016877900.1 | ||||
XM_017022412.1 | Intron | XP_016877901.1 | ||||
XM_017022413.1 | Intron | XP_016877902.1 |
SNAPC5 - small nuclear RNA activating complex polypeptide 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |