Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGGACGAGCAGGAAGCGGCTGT[C/G]AAGTCGGTCAGTAGCTCAGCTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX19A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX19A - DEAD-box helicase 19A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320522.1 | 214 | Silent Mutation | GTC,GTG | V,V 17 | NP_001307451.1 | |
NM_001320525.1 | 214 | UTR 5 | NP_001307454.1 | |||
NM_001320526.1 | 214 | UTR 5 | NP_001307455.1 | |||
NM_001320527.1 | 214 | UTR 5 | NP_001307456.1 | |||
NM_018332.4 | 214 | Silent Mutation | GTC,GTG | V,V 17 | NP_060802.1 |
LOC100506083 - uncharacterized LOC100506083 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |