Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATTTCCCAAACACTCCGCTCCA[C/T]GGTCTCCAGTCATCAGAGCAACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607732 MIM: 611672 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SARM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SARM1 - sterile alpha and TIR motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015077.3 | 3699 | UTR 3 | NP_055892.2 |
SLC46A1 - solute carrier family 46 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242366.2 | 3699 | UTR 3 | NP_001229295.1 | |||
NM_080669.5 | 3699 | UTR 3 | NP_542400.2 | |||
XM_005277786.3 | 3699 | UTR 3 | XP_005277843.1 | |||
XM_017024110.1 | 3699 | UTR 3 | XP_016879599.1 |