Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAAGGAAGCCCTAGTAAGGCAC[A/G]GGGCTGGGAGGCAGGCAGGTTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164731 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKT2 - AKT serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243027.2 | 2613 | UTR 3 | NP_001229956.1 | |||
NM_001243028.2 | 2613 | UTR 3 | NP_001229957.1 | |||
NM_001626.5 | 2613 | UTR 3 | NP_001617.1 | |||
XM_011526614.1 | 2613 | UTR 3 | XP_011524916.1 | |||
XM_011526615.1 | 2613 | UTR 3 | XP_011524917.1 | |||
XM_011526616.1 | 2613 | UTR 3 | XP_011524918.1 | |||
XM_011526618.1 | 2613 | UTR 3 | XP_011524920.1 | |||
XM_011526619.1 | 2613 | UTR 3 | XP_011524921.1 | |||
XM_011526620.1 | 2613 | UTR 3 | XP_011524922.1 | |||
XM_011526622.2 | 2613 | Intron | XP_011524924.1 | |||
XM_017026470.1 | 2613 | UTR 3 | XP_016881959.1 | |||
XM_017026471.1 | 2613 | UTR 3 | XP_016881960.1 |
CNTD2 - cyclin N-terminal domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |