Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCCTATGACACTGTTTGTGACT[A/G]CCGGGGAGTGTGCGGCTGTGTGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600941 MIM: 606214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLVRB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BLVRB - biliverdin reductase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000713.2 | Intron | NP_000704.1 |
SPTBN4 - spectrin beta, non-erythrocytic 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020971.2 | Intron | NP_066022.2 | ||||
NM_025213.2 | Intron | NP_079489.2 | ||||
XM_011527173.2 | Intron | XP_011525475.1 | ||||
XM_017027049.1 | Intron | XP_016882538.1 | ||||
XM_017027050.1 | Intron | XP_016882539.1 | ||||
XM_017027051.1 | Intron | XP_016882540.1 | ||||
XM_017027052.1 | Intron | XP_016882541.1 |