Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTTGCCACTGGCATAGGCATAG[A/G]CCCCGGGCCAGAGGTTGGAGCGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609857 MIM: 607548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMWD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DMWD - dystrophia myotonica, WD repeat containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RSPH6A - radial spoke head 6 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030785.3 | 2066 | Missense Mutation | GCC,GTC | A,V 632 | NP_110412.1 | |
XM_011527351.1 | 2066 | Missense Mutation | GCC,GTC | A,V 632 | XP_011525653.1 |